Product Details
- SNP ID
-
rs148004300
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:23826533 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GACAAAGTGAAGGGCACGCTGTACC[C/T]GCTCATCATTGAGGTCTGCATCATA
- Phenotype
-
MIM: 123856
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CST2
PubMed Links
Gene Details
- Gene
- CST2
- Gene Name
- cystatin SA
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001322.2 |
143 |
Missense Mutation |
CAG,CGG |
Q43R |
NP_001313.1 |
View Full Product Details