Product Details

SNP ID
rs139376338
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:21642867 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGGGGTGTCACTCCTCAGGCAGC[A/G]GCCAGCAATCGGTGACCGGCGTAGA
Phenotype
MIM: 607551
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC116 PubMed Links

Gene Details

Gene
CCDC116
Gene Name
coiled-coil domain containing 116
There are no transcripts associated with this gene.

Gene
SDF2L1
Gene Name
stromal cell derived factor 2 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022044.2 277 Missense Mutation AGC,GGC S65G NP_071327.2

View Full Product Details