Product Details

SNP ID
rs140520049
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:180989255 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTTCATGCTGTATTATAATATT[C/T]GTCTGCTTTCTGCAAGAGCGAGTGT
Phenotype
MIM: 604151
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UBE2E3 PubMed Links

Gene Details

Gene
UBE2E3
Gene Name
ubiquitin conjugating enzyme E2 E3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278554.1 2781 Intron NP_001265483.1
NM_001278555.1 2781 Intron NP_001265484.1
NM_006357.3 2781 Intron NP_006348.1
NM_182678.2 2781 Intron NP_872619.1
XM_005246244.2 2781 Intron XP_005246301.1
XM_011510492.2 2781 Intron XP_011508794.1
XM_017003166.1 2781 UTR 3 XP_016858655.1
XM_017003167.1 2781 Intron XP_016858656.1

View Full Product Details