Product Details

SNP ID
rs143208903
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:48440982 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTCGGCTGAGTTGCAGGGGAAGT[A/G]CCAGAAGCTGGCTCAGGAGTACTCG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPP1R21 PubMed Links

Gene Details

Gene
PPP1R21
Gene Name
protein phosphatase 1 regulatory subunit 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135629.2 214 Missense Mutation TAC,TGC Y10C NP_001129101.1
NM_001193475.1 214 Missense Mutation TAC,TGC Y10C NP_001180404.1
NM_152994.4 214 Missense Mutation TAC,TGC Y10C NP_694539.1
XM_017003301.1 214 UTR 5 XP_016858790.1
XM_017003302.1 214 UTR 5 XP_016858791.1

View Full Product Details