Product Details

SNP ID
rs144643727
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:38781649 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAGAAGCTGATGCATTTGTTCAC[A/G]TCTGGAGACTGCAAAGCATACAGCC
Phenotype
MIM: 607006
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
GEMIN6 PubMed Links

Gene Details

Gene
GEMIN6
Gene Name
gem nuclear organelle associated protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024775.9 377 Silent Mutation ACA,ACG T87T NP_079051.9

View Full Product Details