Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001113397.1 | 1145 | Missense Mutation | NP_001106868.1 | ||
NM_001113398.1 | 1145 | Missense Mutation | NP_001106869.1 | ||
NM_001282725.1 | 1145 | Missense Mutation | NP_001269654.1 | ||
NM_152520.4 | 1145 | Missense Mutation | NP_689733.3 | ||
XM_011510713.2 | 1145 | Missense Mutation | XP_011509015.1 | ||
XM_011510714.2 | 1145 | Missense Mutation | XP_011509016.1 | ||
XM_011510715.2 | 1145 | Missense Mutation | XP_011509017.1 | ||
XM_011510716.2 | 1145 | Missense Mutation | XP_011509018.1 | ||
XM_011510717.2 | 1145 | Missense Mutation | XP_011509019.1 | ||
XM_011510719.2 | 1145 | Missense Mutation | XP_011509021.1 | ||
XM_011510720.2 | 1145 | Missense Mutation | XP_011509022.1 | ||
XM_011510721.2 | 1145 | Missense Mutation | XP_011509023.1 | ||
XM_011510723.2 | 1145 | Intron | XP_011509025.1 | ||
XM_017003435.1 | 1145 | Missense Mutation | XP_016858924.1 | ||
XM_017003436.1 | 1145 | Missense Mutation | XP_016858925.1 | ||
XM_017003437.1 | 1145 | Missense Mutation | XP_016858926.1 | ||
XM_017003438.1 | 1145 | Missense Mutation | XP_016858927.1 | ||
XM_017003439.1 | 1145 | Missense Mutation | XP_016858928.1 | ||
XM_017003440.1 | 1145 | Missense Mutation | XP_016858929.1 | ||
XM_017003441.1 | 1145 | Intron | XP_016858930.1 |