Product Details

SNP ID
rs148820734
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:84906116 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAGAACACCCTGCCGACCAAAGA[A/G]AGTGAGTGTGCCTCGGTCTCCCGCG
Phenotype
MIM: 188399
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMSB10 PubMed Links

Gene Details

Gene
TMSB10
Gene Name
thymosin beta 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021103.3 Intron NP_066926.1

View Full Product Details