Product Details

SNP ID
rs141391181
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:128573545 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGGGCCCAAACTCGGTCCCCAGA[C/T]CCCCGAGTGTCTCTGGTGCCAGAGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC01565 PubMed Links

Gene Details

Gene
LINC01565
Gene Name
long intergenic non-protein coding RNA 1565
There are no transcripts associated with this gene.

View Full Product Details