Product Details

SNP ID
rs148842396
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:186539524 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGGAGTTGCAGCGACTTAGGTAT[A/G]TGTGGAAATCTGCACAGTCGCAATC
Phenotype
MIM: 123730
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CRYGS PubMed Links

Gene Details

Gene
CRYGS
Gene Name
crystallin gamma S
There are no transcripts associated with this gene.

Gene
TBCCD1
Gene Name
TBCC domain containing 1
There are no transcripts associated with this gene.

View Full Product Details