Product Details

SNP ID
rs144483811
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:74748141 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTTCTTTCTTTTACGACGTTTC[C/T]GAAGAGGGCTTGGAAAATACGTGTT
Phenotype
MIM: 600345
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BTC PubMed Links

Gene Details

Gene
BTC
Gene Name
betacellulin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316963.1 651 Missense Mutation CAG,CGG Q97R NP_001303892.1
NM_001729.3 651 Missense Mutation CAG,CGG Q146R NP_001720.1
XM_011532211.1 651 Missense Mutation CAG,CGG Q146R XP_011530513.1
XM_017008568.1 651 Intron XP_016864057.1

View Full Product Details