Product Details

SNP ID
rs145681416
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:112539663 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTTCAGCTGTGGTTCATACTGGT[C/T]TGCATGTTGCAATCCATCTTCCCTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZGRF1 PubMed Links

Gene Details

Gene
ZGRF1
Gene Name
zinc finger GRF-type containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018392.4 6359 Missense Mutation AAC,GAC N2067D NP_060862.3
XM_005263115.3 6359 Missense Mutation AAC,GAC N2067D XP_005263172.1
XM_011532091.2 6359 Missense Mutation AAC,GAC N2050D XP_011530393.1
XM_011532092.2 6359 Missense Mutation AAC,GAC N2047D XP_011530394.1
XM_011532093.2 6359 Missense Mutation AAC,GAC N2009D XP_011530395.1
XM_011532094.2 6359 Missense Mutation AAC,GAC N1981D XP_011530396.1
XM_011532096.2 6359 Intron XP_011530398.1
XM_011532097.2 6359 Missense Mutation AAC,GAC N1116D XP_011530399.1
XM_011532098.2 6359 Missense Mutation AAC,GAC N1024D XP_011530400.1
XM_011532099.2 6359 Missense Mutation AAC,GAC N1024D XP_011530401.1
XM_017008369.1 6359 Missense Mutation AAC,GAC N2047D XP_016863858.1
XM_017008370.1 6359 Missense Mutation AAC,GAC N1992D XP_016863859.1
XM_017008371.1 6359 Missense Mutation AAC,GAC N1972D XP_016863860.1
XM_017008372.1 6359 Missense Mutation AAC,GAC N817D XP_016863861.1
XM_017008373.1 6359 Missense Mutation AAC,GAC N817D XP_016863862.1

View Full Product Details