Product Details

SNP ID
rs146074949
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:77160889 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGGAAAAAATAATTTCAGAAAAA[C/T]TGCACTATGAATTGGAAGCTACTAC
Phenotype
MIM: 603203
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCNG2 PubMed Links

Gene Details

Gene
CCNG2
Gene Name
cyclin G2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004354.2 888 Silent Mutation CTG,TTG L149L NP_004345.1
XM_011532398.1 888 Silent Mutation CTG,TTG L149L XP_011530700.1
XM_011532399.2 888 Silent Mutation CTG,TTG L149L XP_011530701.1

View Full Product Details