Product Details

SNP ID
rs147931838
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:173528999 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTGGCGGTGCCTCGGCGCTTCAC[C/T]GGGCGCGGCCCCCCCAGGCCCGGGG
Phenotype
MIM: 602407
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HAND2 PubMed Links

Gene Details

Gene
HAND2
Gene Name
heart and neural crest derivatives expressed 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021973.2 1229 Silent Mutation CCA,CCG P97P NP_068808.1
Gene
HAND2-AS1
Gene Name
HAND2 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details