Product Details

SNP ID
rs151251968
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:87479514 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTCACACTCCTCAAAGAAACGGG[C/T]TATGCAGTGTTCCATGGGCACCAGA
Phenotype
MIM: 606041
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SPARCL1 PubMed Links

Gene Details

Gene
SPARCL1
Gene Name
SPARC like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128310.2 2112 Missense Mutation ACC,GCC T628A NP_001121782.1
NM_001291976.1 2112 Missense Mutation ACC,GCC T503A NP_001278905.1
NM_001291977.1 2112 Missense Mutation ACC,GCC T503A NP_001278906.1
NM_004684.5 2112 Missense Mutation ACC,GCC T628A NP_004675.3

View Full Product Details