Product Details

SNP ID
rs147492888
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:138556500 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTCAGTGATGCCTGCTGAAGAGA[A/G]GATGCTGCCTGTCTAATATTTTCTC
Phenotype
MIM: 600548
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HSPA9 PubMed Links

Gene Details

Gene
HSPA9
Gene Name
heat shock protein family A (Hsp70) member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004134.6 2225 Silent Mutation TCC,TCT S638S NP_004125.3
Gene
SNORD63
Gene Name
small nucleolar RNA, C/D box 63
There are no transcripts associated with this gene.

View Full Product Details