Product Details

SNP ID
rs41342846
Assay Type
Functionally Tested
NCBI dbSNP Submissions
24
Location
Chr.6:32394925 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGTCTTTCCTTCCATGTCCCTCCA[T/C]GGCACGTGGGGCTGTGGGAACCACC
Phenotype
MIM: 606000
Polymorphism
T/C, Transition Substitution
Allele Nomenclature
Literature Links
BTNL2 PubMed Links

Gene Details

Gene
BTNL2
Gene Name
butyrophilin like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304561.1 2935 Silent Mutation CCA,CCG P393P NP_001291490.1
XM_017011057.1 2935 Silent Mutation CCA,CCG P393P XP_016866546.1
Gene
HCG23
Gene Name
HLA complex group 23 (non-protein coding)
There are no transcripts associated with this gene.

Gene
LOC101929163
Gene Name
uncharacterized LOC101929163
There are no transcripts associated with this gene.

View Full Product Details