Product Details

SNP ID
rs148334138
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:73251613 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTATACTGGAAAATATAAACATAAA[A/G]AGAAATGTTTCAATGTCTTCTATTT
Phenotype
MIM: 611688
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KHDC1 PubMed Links

Gene Details

Gene
KHDC1
Gene Name
KH homology domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001251874.1 Intron NP_001238803.1
NM_030568.4 Intron NP_085045.3

View Full Product Details