Product Details

SNP ID
rs146476912
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:6165299 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATGGATTTCATCCACTCCTCCTT[C/T]TCCTCCGGGCTCGGGGCTGAGATCC
Phenotype
MIM: 605081
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYTH3 PubMed Links

Gene Details

Gene
CYTH3
Gene Name
cytohesin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004227.3 1205 Silent Mutation GAA,GAG E367E NP_004218.1
Gene
USP42
Gene Name
ubiquitin specific peptidase 42
There are no transcripts associated with this gene.

View Full Product Details