Product Details

SNP ID
rs147647748
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:44114821 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCCAGGCCTCCCAGGTCATCGTC[C/G]TCTGCCCCGAAGCCCGAGAAGCTGA
Phenotype
MIM: 602981 MIM: 600815
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
AEBP1 PubMed Links

Gene Details

Gene
AEBP1
Gene Name
AE binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001129.4 2024 Intron NP_001120.3
XM_011515162.1 2024 Intron XP_011513464.1
Gene
MIR4649
Gene Name
microRNA 4649
There are no transcripts associated with this gene.

Gene
POLD2
Gene Name
DAN polymerase delta 2, accessory subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127218.2 2024 Missense Mutation GAC,GAG D458E NP_001120690.1
NM_001256879.1 2024 Missense Mutation GAC,GAG D458E NP_001243808.1
NM_006230.3 2024 Missense Mutation GAC,GAG D493E NP_006221.2
XM_011515431.1 2024 Missense Mutation GAC,GAG D491E XP_011513733.1

View Full Product Details