Product Details

SNP ID
rs147781497
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:44115298 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCCCAGAAGACAAAAATTACCTC[C/G]GATGATTTTGGAGCCAAAGCTGGGG
Phenotype
MIM: 602981 MIM: 600815
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
AEBP1 PubMed Links

Gene Details

Gene
AEBP1
Gene Name
AE binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001129.4 1896 Intron NP_001120.3
XM_011515162.1 1896 Intron XP_011513464.1
Gene
MIR4649
Gene Name
microRNA 4649
There are no transcripts associated with this gene.

Gene
POLD2
Gene Name
DAN polymerase delta 2, accessory subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127218.2 1896 Missense Mutation CGA,GGA R416G NP_001120690.1
NM_001256879.1 1896 Missense Mutation CGA,GGA R416G NP_001243808.1
NM_006230.3 1896 Missense Mutation CGA,GGA R451G NP_006221.2
XM_011515431.1 1896 Missense Mutation CGA,GGA R449G XP_011513733.1

View Full Product Details