Product Details

SNP ID
rs148892911
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:27832953 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGTGTTCTGTGACCATTCTTAGC[A/G]TGATACTTTATGCCATTCACATTCT
Phenotype
MIM: 606246 MIM: 605326
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
JAZF1 PubMed Links

Gene Details

Gene
JAZF1
Gene Name
JAZF zinc finger 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175061.3 520 Silent Mutation CAC,CAT H193H NP_778231.2
XM_006715656.1 520 Silent Mutation CAC,CAT H129H XP_006715719.1
XM_017011831.1 520 Silent Mutation CAC,CAT H129H XP_016867320.1
XM_017011832.1 520 Silent Mutation CAC,CAT H129H XP_016867321.1
Gene
TAX1BP1
Gene Name
Tax1 binding protein 1
There are no transcripts associated with this gene.

View Full Product Details