Product Details

SNP ID
rs141560064
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:102651386 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGGGGTACCACAAACATGACAG[C/T]GCCTTTGGGGACTTGTGTGCCCATG
Phenotype
MIM: 601878
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLF10 PubMed Links

Gene Details

Gene
KLF10
Gene Name
Kruppel like factor 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001032282.3 1247 Missense Mutation ACT,GCT T305A NP_001027453.1
NM_005655.3 1247 Missense Mutation ACT,GCT T316A NP_005646.1
Gene
LOC101927245
Gene Name
vegetative cell wall protein gp1-like
There are no transcripts associated with this gene.

View Full Product Details