Product Details

SNP ID
rs142325296
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:17247368 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAGTCCCTCCGGAACTCACACTCC[A/T]GGTGACTGGTCGCTGCCTCTCCACC
Phenotype
MIM: 604913 MIM: 609927
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CNOT7 PubMed Links

Gene Details

Gene
CNOT7
Gene Name
CCR4-NOT transcription complex subunit 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322087.1 442 Intron NP_001309016.1
NM_001322088.1 442 Intron NP_001309017.1
NM_001322089.1 442 Intron NP_001309018.1
NM_001322090.1 442 Intron NP_001309019.1
NM_001322091.1 442 Intron NP_001309020.1
NM_001322092.1 442 Intron NP_001309021.1
NM_001322093.1 442 Intron NP_001309022.1
NM_001322094.1 442 Intron NP_001309023.1
NM_001322095.1 442 Intron NP_001309024.1
NM_001322096.1 442 Intron NP_001309025.1
NM_001322097.1 442 Intron NP_001309026.1
NM_001322098.1 442 Intron NP_001309027.1
NM_001322099.1 442 Intron NP_001309028.1
NM_001322100.1 442 Intron NP_001309029.1
NM_013354.6 442 Intron NP_037486.2
NM_054026.3 442 Intron NP_473367.2
XM_005273481.2 442 Intron XP_005273538.1
Gene
VPS37A
Gene Name
VPS37A, ESCRT-I subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145152.1 442 Nonsense Mutation AGA,TGA R42* NP_001138624.1
NM_152415.2 442 Missense Mutation AGT,TGT S42C NP_689628.2
XM_005273400.2 442 UTR 5 XP_005273457.1
XM_005273401.2 442 UTR 5 XP_005273458.1
XM_006716286.1 442 Missense Mutation AGT,TGT S42C XP_006716349.1
XM_017013021.1 442 Missense Mutation AGT,TGT S42C XP_016868510.1
XM_017013022.1 442 Missense Mutation AGT,TGT S42C XP_016868511.1
XM_017013023.1 442 UTR 5 XP_016868512.1
XM_017013024.1 442 UTR 5 XP_016868513.1
XM_017013025.1 442 UTR 5 XP_016868514.1
XM_017013026.1 442 Intron XP_016868515.1

View Full Product Details