Product Details
- SNP ID
-
rs145491168
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:52113329 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACATATGCTTCAAAATTCTGCTCAC[C/T]GATAGGTCCCTAAATGGAGACAAAA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ST18
PubMed Links
Gene Details
- Gene
- ST18
- Gene Name
- ST18, C2H2C-type zinc finger
View Full Product Details