Product Details
- SNP ID
-
rs145972852
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:30836916 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGAAAAAATTGGCTGCCGAAAATTA[A/G]AAGCAGGTATTTGAGATTGAAAATA
- Phenotype
-
MIM: 605795
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TEX15
PubMed Links
Gene Details
- Gene
- TEX15
- Gene Name
- testis expressed 15
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_031271.3 |
9149 |
Missense Mutation |
TCT,TTT |
S2740F |
NP_112561.2 |
XM_005273575.3 |
9149 |
Missense Mutation |
TCT,TTT |
S3123F |
XP_005273632.1 |
XM_005273576.3 |
9149 |
Missense Mutation |
TCT,TTT |
S3020F |
XP_005273633.1 |
XM_006716369.3 |
9149 |
Missense Mutation |
TCT,TTT |
S3123F |
XP_006716432.1 |
XM_011544588.2 |
9149 |
Missense Mutation |
TCT,TTT |
S3127F |
XP_011542890.1 |
XM_011544589.2 |
9149 |
Missense Mutation |
TCT,TTT |
S3123F |
XP_011542891.1 |
XM_011544590.2 |
9149 |
Missense Mutation |
TCT,TTT |
S3123F |
XP_011542892.1 |
XM_011544591.2 |
9149 |
Missense Mutation |
TCT,TTT |
S3123F |
XP_011542893.1 |
XM_011544592.2 |
9149 |
Missense Mutation |
TCT,TTT |
S3123F |
XP_011542894.1 |
XM_011544593.2 |
9149 |
Missense Mutation |
TCT,TTT |
S3020F |
XP_011542895.1 |
XM_011544594.2 |
9149 |
Intron |
|
|
XP_011542896.1 |
View Full Product Details