Product Details
- SNP ID
-
rs141308173
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:131580322 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGAACTGCTGCCACCGCTTGGAGAA[A/G]TTCACTTTCCCGTCGATGTAGTCTG
- Phenotype
-
MIM: 600303
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RAPGEF1
PubMed Links
Gene Details
- Gene
- RAPGEF1
- Gene Name
- Rap guanine nucleotide exchange factor 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001304275.1 |
3237 |
Silent Mutation |
AAC,AAT |
N1025N |
NP_001291204.1 |
NM_005312.3 |
3237 |
Silent Mutation |
AAC,AAT |
N1008N |
NP_005303.2 |
NM_198679.1 |
3237 |
Silent Mutation |
AAC,AAT |
N1026N |
NP_941372.1 |
XM_005272186.4 |
3237 |
Silent Mutation |
AAC,AAT |
N1195N |
XP_005272243.1 |
XM_005272191.3 |
3237 |
Silent Mutation |
AAC,AAT |
N1157N |
XP_005272248.1 |
XM_006717067.3 |
3237 |
Silent Mutation |
AAC,AAT |
N1201N |
XP_006717130.1 |
XM_006717072.3 |
3237 |
Silent Mutation |
AAC,AAT |
N1163N |
XP_006717135.1 |
XM_006717074.3 |
3237 |
Silent Mutation |
AAC,AAT |
N1031N |
XP_006717137.1 |
XM_011518569.2 |
3237 |
Silent Mutation |
AAC,AAT |
N1209N |
XP_011516871.1 |
XM_011518570.2 |
3237 |
Silent Mutation |
AAC,AAT |
N1208N |
XP_011516872.1 |
XM_011518571.2 |
3237 |
Silent Mutation |
AAC,AAT |
N1204N |
XP_011516873.1 |
XM_011518572.2 |
3237 |
Silent Mutation |
AAC,AAT |
N1203N |
XP_011516874.1 |
XM_011518573.2 |
3237 |
Silent Mutation |
AAC,AAT |
N1186N |
XP_011516875.1 |
XM_011518574.2 |
3237 |
Silent Mutation |
AAC,AAT |
N1178N |
XP_011516876.1 |
XM_011518575.2 |
3237 |
Silent Mutation |
AAC,AAT |
N1177N |
XP_011516877.1 |
XM_011518576.2 |
3237 |
Silent Mutation |
AAC,AAT |
N1092N |
XP_011516878.1 |
XM_011518577.2 |
3237 |
Silent Mutation |
AAC,AAT |
N1091N |
XP_011516879.1 |
XM_011518578.2 |
3237 |
Silent Mutation |
AAC,AAT |
N1070N |
XP_011516880.1 |
XM_011518579.2 |
3237 |
Silent Mutation |
AAC,AAT |
N1039N |
XP_011516881.1 |
XM_011518580.2 |
3237 |
Intron |
|
|
XP_011516882.1 |
XM_011518581.2 |
3237 |
Intron |
|
|
XP_011516883.1 |
XM_011518582.2 |
3237 |
Intron |
|
|
XP_011516884.1 |
XM_017014633.1 |
3237 |
Silent Mutation |
AAC,AAT |
N1178N |
XP_016870122.1 |
XM_017014634.1 |
3237 |
Silent Mutation |
AAC,AAT |
N1172N |
XP_016870123.1 |
XM_017014635.1 |
3237 |
Silent Mutation |
AAC,AAT |
N1172N |
XP_016870124.1 |
XM_017014636.1 |
3237 |
Silent Mutation |
AAC,AAT |
N1164N |
XP_016870125.1 |
XM_017014637.1 |
3237 |
Silent Mutation |
AAC,AAT |
N1156N |
XP_016870126.1 |
XM_017014638.1 |
3237 |
Silent Mutation |
AAC,AAT |
N1125N |
XP_016870127.1 |
XM_017014639.1 |
3237 |
Silent Mutation |
AAC,AAT |
N1123N |
XP_016870128.1 |
XM_017014640.1 |
3237 |
Intron |
|
|
XP_016870129.1 |
XM_017014641.1 |
3237 |
Intron |
|
|
XP_016870130.1 |
View Full Product Details