Product Details
- SNP ID
-
rs148928350
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:21077207 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTCAAAATCTTCTAGTGTCCTTTC[A/T]TATGCAGTACATTAGCCATCAGTCA
- Phenotype
-
MIM: 147640
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
IFNB1
PubMed Links
Gene Details
- Gene
- IFNB1
- Gene Name
- interferon beta 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002176.3 |
757 |
UTR 3 |
|
|
NP_002167.1 |
View Full Product Details