Product Details

SNP ID
rs150203467
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:35074136 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGTCACAAGACTTTGGCAGAGAT[A/G]TCCGAAATTCTTCAAGGAAGGCGTC
Phenotype
MIM: 602956 MIM: 601023
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FANCG PubMed Links

Gene Details

Gene
FANCG
Gene Name
Fanconi anemia complementation group G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004629.1 2333 Missense Mutation ACA,ATA T614I NP_004620.1
Gene
VCP
Gene Name
valosin containing protein
There are no transcripts associated with this gene.

View Full Product Details