Product Details

SNP ID
rs138085112
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:135287201 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTTTCTGGTGTCTAAGAAGGCTC[A/G]ATCGCCTACTAAAGTTTCTCTTGCA
Phenotype
MIM: 314997
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF75D PubMed Links

Gene Details

Gene
ZNF75D
Gene Name
zinc finger protein 75D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185063.2 3308 Intron NP_001171992.1
NM_007131.4 3308 Missense Mutation TCG,TTG S490L NP_009062.2
XM_005262469.1 3308 Missense Mutation TCG,TTG S490L XP_005262526.1
XM_005262471.3 3308 Missense Mutation TCG,TTG S283L XP_005262528.1
XM_011531391.1 3308 Missense Mutation TCG,TTG S490L XP_011529693.1
XM_011531395.2 3308 Missense Mutation TCG,TTG S340L XP_011529697.1
XM_011531396.2 3308 Missense Mutation TCG,TTG S322L XP_011529698.1
XM_011531397.2 3308 Missense Mutation TCG,TTG S319L XP_011529699.1
XM_017029818.1 3308 Missense Mutation TCG,TTG S344L XP_016885307.1
XM_017029819.1 3308 Missense Mutation TCG,TTG S356L XP_016885308.1
XM_017029820.1 3308 Missense Mutation TCG,TTG S319L XP_016885309.1

View Full Product Details