Product Details

SNP ID
rs138137225
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:48923252 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGGTATTCCTGTTGGGACACTGC[A/C]AGCACCGAAGCTAGGATCTCATCAT
Phenotype
MIM: 300713 MIM: 300295
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OTUD5 PubMed Links

Gene Details

Gene
OTUD5
Gene Name
OTU deubiquitinase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136157.1 2449 Silent Mutation CTG,CTT L541L NP_001129629.1
NM_001136158.1 2449 Silent Mutation CTG,CTT L541L NP_001129630.1
NM_001136159.1 2449 Silent Mutation CTG,CTT L324L NP_001129631.1
NM_017602.3 2449 Silent Mutation CTG,CTT L546L NP_060072.1
XM_005272622.3 2449 Silent Mutation CTG,CTT L542L XP_005272679.1
XM_005278039.3 2449 Silent Mutation CTG,CTT L547L XP_005278096.1
XM_005278042.4 2449 Silent Mutation CTG,CTT L330L XP_005278099.1
XM_006724537.2 2449 Silent Mutation CTG,CTT L546L XP_006724600.1
XM_011543932.1 2449 Silent Mutation CTG,CTT L547L XP_011542234.1
XM_011543934.2 2449 Silent Mutation CTG,CTT L309L XP_011542236.1
XM_017029628.1 2449 Silent Mutation CTG,CTT L542L XP_016885117.1
XM_017029629.1 2449 Silent Mutation CTG,CTT L330L XP_016885118.1
XM_017029630.1 2449 Silent Mutation CTG,CTT L329L XP_016885119.1
Gene
PIM2
Gene Name
Pim-2 proto-oncogene, serine/threonine kinase
There are no transcripts associated with this gene.

View Full Product Details