Product Details

SNP ID
rs139515883
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:101411988 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCCACCAAGAGCCCAACAATCAC[C/T]ATGATGCTGAGCACGGAAGGCAGGG
Phenotype
MIM: 300644 MIM: 300610
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARMCX4 PubMed Links

Gene Details

Gene
ARMCX4
Gene Name
armadillo repeat containing, X-linked 4
There are no transcripts associated with this gene.

Gene
GLA
Gene Name
galactosidase alpha
There are no transcripts associated with this gene.

Gene
HNRNPH2
Gene Name
heterogeneous nuclear ribonucleoprotein H2 (H')
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001032393.2 384 Intron NP_001027565.1
NM_019597.4 384 Intron NP_062543.1
Gene
RPL36A-HNRNPH2
Gene Name
RPL36A-HNRNPH2 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199973.1 384 Missense Mutation CCA,CTA P154L NP_001186902.1
NM_001199974.1 384 Missense Mutation CCA,CTA P113L NP_001186903.1

View Full Product Details