Product Details

SNP ID
rs143516807
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:106204864 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACACATGTTAAGTTTGGAGACCC[A/G]CTGCTATAAGTATGACATTAGGGAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MUM1L1 PubMed Links

Gene Details

Gene
MUM1L1
Gene Name
MUM1 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171020.1 Intron NP_001164491.1
NM_152423.4 Intron NP_689636.3
XM_005262077.2 Intron XP_005262134.1
XM_011530856.1 Intron XP_011529158.1
XM_011530857.1 Intron XP_011529159.1
XM_017029269.1 Intron XP_016884758.1

View Full Product Details