Product Details

SNP ID
rs145406974
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:77971698 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATTTCTGTTGAGGGTATGACTTG[C/T]AATTCCTGTGTTTGGACCATTGAGC
Phenotype
MIM: 300011 MIM: 300567
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP7A PubMed Links

Gene Details

Gene
ATP7A
Gene Name
ATPase copper transporting alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000052.6 258 Silent Mutation TGC,TGT C19C NP_000043.4
NM_001282224.1 258 Silent Mutation TGC,TGT C19C NP_001269153.1
Gene
PGAM4
Gene Name
phosphoglycerate mutase family member 4
There are no transcripts associated with this gene.

View Full Product Details