Product Details
- SNP ID
-
rs147389956
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:53940267 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGGCCATAGGTGTGGTGCTGCGGT[C/T]TGCCTGGGCCATGCCAAAGGCATTG
- Phenotype
-
MIM: 300560
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PHF8
PubMed Links
Gene Details
- Gene
- PHF8
- Gene Name
- PHD finger protein 8
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001184896.1 |
3702 |
Missense Mutation |
AAC,GAC |
N1003D |
NP_001171825.1 |
NM_001184897.1 |
3702 |
Missense Mutation |
AAC,GAC |
N866D |
NP_001171826.1 |
NM_001184898.1 |
3702 |
Intron |
|
|
NP_001171827.1 |
NM_015107.2 |
3702 |
Missense Mutation |
AAC,GAC |
N967D |
NP_055922.1 |
XM_005261996.1 |
3702 |
Missense Mutation |
AAC,GAC |
N1003D |
XP_005262053.1 |
XM_005261997.3 |
3702 |
Missense Mutation |
AAC,GAC |
N967D |
XP_005262054.1 |
XM_005261999.1 |
3702 |
Missense Mutation |
AAC,GAC |
N967D |
XP_005262056.1 |
XM_005262000.1 |
3702 |
Missense Mutation |
AAC,GAC |
N902D |
XP_005262057.1 |
XM_011530778.1 |
3702 |
Missense Mutation |
AAC,GAC |
N1003D |
XP_011529080.1 |
XM_017029361.1 |
3702 |
Missense Mutation |
AAC,GAC |
N967D |
XP_016884850.1 |
XM_017029362.1 |
3702 |
Missense Mutation |
AAC,GAC |
N967D |
XP_016884851.1 |
View Full Product Details