Product Details

SNP ID
rs151274585
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.X:77943272 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTGAAAATTAAAGAATCACATTC[C/T]GCTAATTATTTTAGTGCCCAGCACT
Phenotype
MIM: 300011
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ATP7A PubMed Links

Gene Details

Gene
ATP7A
Gene Name
ATPase copper transporting alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000052.6 Intron NP_000043.4
NM_001282224.1 Intron NP_001269153.1

View Full Product Details