Product Details

SNP ID
rs1061695
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:66994269 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAACGCACTGATGAGATCTTCCTT[T/C]TTGGCAACCTGACCTTTAGAAACAT
Phenotype
MIM: 607444 MIM: 611243
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
SBDS PubMed Links

Gene Details

Gene
SBDS
Gene Name
SBDS ribosome assembly guanine nucleotide exchange factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016038.2 385 Silent Mutation NP_057122.2
Gene
TYW1
Gene Name
tRNA-yW synthesizing protein 1 homolog
There are no transcripts associated with this gene.

View Full Product Details