Product Details

SNP ID
rs11542320
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:23559142 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAGGCTTACCTGGATGGGAAGGTG[T/G]GGGCCATCAGGGGGTCCAGGGGCTG
Phenotype
MIM: 600277
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
ID3 PubMed Links

Gene Details

Gene
ID3
Gene Name
inhibitor of DNA binding 3, HLH protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002167.4 653 Silent Mutation CCA,CCC P95P NP_002158.3

View Full Product Details