Product Details

SNP ID
rs1155352
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:16669786 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTATACATAAAACACATACACACT[C/T]TCTCTCTCTCTCTTAAAATATGACT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRD62P1-PARP4P3 PubMed Links

Gene Details

Gene
ANKRD62P1-PARP4P3
Gene Name
ANKRD62P1-PARP4P3 readthrough, transcribed pseudogene
There are no transcripts associated with this gene.

View Full Product Details