Product Details

SNP ID
rs181389475
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:43952977 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCTCACCACAGGTCTGGGCCCTC[A/C]GACCTGCGGAAGCGAAGGGGAAGGA
Phenotype
MIM: 167780 MIM: 600781
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PPY PubMed Links

Gene Details

Gene
PPY
Gene Name
pancreatic polypeptide
There are no transcripts associated with this gene.

Gene
PYY
Gene Name
peptide YY
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004160.5 1424 Intron NP_004151.3
XM_011525035.1 1424 Silent Mutation TCG,TCT S91S XP_011523337.1

View Full Product Details