Product Details

SNP ID
rs182179695
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:106079497 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGTCTGATGATAAGGAAATTAGTT[A/G]CAGAGAAATTGAATATCCCTTGGAA
Phenotype
MIM: 607756
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AASDHPPT PubMed Links

Gene Details

Gene
AASDHPPT
Gene Name
aminoadipate-semialdehyde dehydrogenase-phosphopantetheinyl transferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015423.2 360 Missense Mutation ACA,GCA T72A NP_056238.2
Gene
KBTBD3
Gene Name
kelch repeat and BTB domain containing 3
There are no transcripts associated with this gene.

View Full Product Details