Product Details

SNP ID
rs181956190
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:44662324 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTCTTAAAACTTTCAGAGTGAAG[A/G]CAATGTAGACCTAAGGCAGACCTAT
Phenotype
MIM: 616934
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MEIOC PubMed Links

Gene Details

Gene
MEIOC
Gene Name
meiosis specific with coiled-coil domain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145080.2 409 Missense Mutation GAC,GGC D71G NP_001138552.2
XM_005257236.3 409 Missense Mutation GAC,GGC D71G XP_005257293.1

View Full Product Details