Product Details

SNP ID
rs182459679
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:70301415 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCATGTCAAACTTGATGAGCGGC[A/G]GGGCGATCACGCGCACCTCGGCGTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRRC20 PubMed Links

Gene Details

Gene
LRRC20
Gene Name
leucine rich repeat containing 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278211.1 654 Missense Mutation CCG,CTG P165L NP_001265140.1
NM_001278212.1 654 Missense Mutation CCG,CTG P165L NP_001265141.1
NM_001278213.1 654 Missense Mutation CCG,CTG P115L NP_001265142.1
NM_001278214.1 654 Missense Mutation CCG,CTG P96L NP_001265143.1
NM_018205.3 654 Missense Mutation CCG,CTG P109L NP_060675.1
NM_018239.3 654 Missense Mutation CCG,CTG P115L NP_060709.2
NM_207119.2 654 Missense Mutation CCG,CTG P165L NP_997002.1
XM_017016374.1 654 Missense Mutation CCG,CTG P165L XP_016871863.1
XM_017016375.1 654 Missense Mutation CCG,CTG P165L XP_016871864.1

View Full Product Details