Product Details
- SNP ID
-
rs183760285
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:184571363 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGGTAACATCTGAGTTGGCGTGGA[C/T]ATCTCATCCAGAAAGTGGGGTGAGG
- Phenotype
-
MIM: 601839
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
EPHB3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs13069661] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EPHB3
- Gene Name
- EPH receptor B3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004443.3 |
601 |
Missense Mutation |
ACA,ATA |
T55I |
NP_004434.2 |
View Full Product Details