Product Details

SNP ID
rs184060663
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:42315071 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGACACGACACTTCTCCGCTGAT[G/T]AGTTTGGTGAGCACGTGGGGACCTA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PRR19 PubMed Links

Gene Details

Gene
PRR19
Gene Name
proline rich 19
There are no transcripts associated with this gene.

Gene
TMEM145
Gene Name
transmembrane protein 145
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173633.2 593 Nonsense Mutation GAG,TAG E167* NP_775904.2
XM_005258781.4 593 Nonsense Mutation GAG,TAG E181* XP_005258838.1
XM_011526791.2 593 Nonsense Mutation GAG,TAG E167* XP_011525093.1
XM_011526792.1 593 Nonsense Mutation GAG,TAG E181* XP_011525094.1

View Full Product Details