Product Details

SNP ID
rs185038174
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:156244736 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGGCATCTGCCCCTGCCTCTTCC[C/T]GGGCCGGGCCAGGCGTGCCCTCACC
Phenotype
MIM: 112260 MIM: 614579 MIM: 609176 MIM: 610962
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BGLAP PubMed Links

Gene Details

Gene
BGLAP
Gene Name
bone gamma-carboxyglutamate protein
There are no transcripts associated with this gene.

Gene
PAQR6
Gene Name
progestin and adipoQ receptor family member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272104.1 908 Intron NP_001259033.1
NM_001272105.1 908 Intron NP_001259034.1
NM_001272106.1 908 Missense Mutation CAG,CGG Q88R NP_001259035.1
NM_001272107.1 908 Intron NP_001259036.1
NM_001272108.1 908 Intron NP_001259037.1
NM_001272109.1 908 Intron NP_001259038.1
NM_001272110.1 908 Intron NP_001259039.1
NM_001272111.1 908 Intron NP_001259040.1
NM_001272112.1 908 Intron NP_001259041.1
NM_001272113.1 908 Intron NP_001259042.1
NM_024897.3 908 Missense Mutation CAG,CGG Q156R NP_079173.2
NM_198406.2 908 Intron NP_940798.1
XM_005245494.3 908 Intron XP_005245551.1
XM_006711546.2 908 Missense Mutation CAG,CGG Q262R XP_006711609.1
XM_006711547.3 908 Missense Mutation CAG,CGG Q262R XP_006711610.1
XM_006711548.3 908 Missense Mutation CAG,CGG Q238R XP_006711611.1
XM_006711552.2 908 Missense Mutation CAG,CGG Q151R XP_006711615.1
XM_006711553.3 908 Missense Mutation CAG,CGG Q151R XP_006711616.1
XM_011510000.2 908 Intron XP_011508302.1
XM_017002385.1 908 Missense Mutation CAG,CGG Q262R XP_016857874.1
Gene
PMF1
Gene Name
polyamine-modulated factor 1
There are no transcripts associated with this gene.

Gene
PMF1-BGLAP
Gene Name
PMF1-BGLAP readthrough
There are no transcripts associated with this gene.

Gene
SMG5
Gene Name
SMG5, nonsense mediated mRNA decay factor
There are no transcripts associated with this gene.

View Full Product Details