Product Details

SNP ID
rs184806294
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.13:102730133 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGTGGGACTTACAGGAAGGTGGT[G/T]TAATAATTAAGGTTTCCTTTCTGCA
Phenotype
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
CCDC168 PubMed Links

Gene Details

Gene
CCDC168
Gene Name
coiled-coil domain containing 168
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146197.1 20704 Missense Mutation NP_001139669.1
XM_011521106.1 20704 Missense Mutation XP_011519408.1

View Full Product Details