Product Details

SNP ID
rs185282123
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11343109 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTGGCCAGCGTAAGGCTCAGGAA[C/G]AGCAGGTAAAGCTGGCAGGGAGAGC
Phenotype
MIM: 604350 MIM: 613771
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCDC159 PubMed Links

Gene Details

Gene
CCDC159
Gene Name
coiled-coil domain containing 159
There are no transcripts associated with this gene.

Gene
RAB3D
Gene Name
RAB3D, member RAS oncogene family
There are no transcripts associated with this gene.

Gene
TMEM205
Gene Name
transmembrane protein 205
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145416.1 274 Silent Mutation CTC,CTG L92L NP_001138888.1
NM_001321112.1 274 Silent Mutation CTC,CTG L92L NP_001308041.1
NM_001321113.1 274 Silent Mutation CTC,CTG L92L NP_001308042.1
NM_001321114.1 274 Silent Mutation CTC,CTG L92L NP_001308043.1
NM_033408.3 274 Silent Mutation CTC,CTG L92L NP_212133.1
NM_198536.2 274 Silent Mutation CTC,CTG L92L NP_940938.1
XM_005259899.2 274 Silent Mutation CTC,CTG L92L XP_005259956.1
XM_017026769.1 274 Missense Mutation CTT,GTT L86V XP_016882258.1

View Full Product Details