Product Details
- SNP ID
-
rs186669756
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:101656420 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGATTTCTGAAGCAAGGCAAGGACC[C/T]TCCTGAGATCGCGGACACCCAGAAT
- Phenotype
-
MIM: 300363
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ARMCX2
PubMed Links
Gene Details
- Gene
- ARMCX2
- Gene Name
- armadillo repeat containing, X-linked 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001282231.1 |
1752 |
Missense Mutation |
AAG,AGG |
K390R |
NP_001269160.1 |
NM_014782.6 |
1752 |
Missense Mutation |
AAG,AGG |
K390R |
NP_055597.1 |
NM_177949.3 |
1752 |
Missense Mutation |
AAG,AGG |
K390R |
NP_808818.1 |
XM_005278109.1 |
1752 |
Missense Mutation |
AAG,AGG |
K390R |
XP_005278166.1 |
XM_005278110.1 |
1752 |
Missense Mutation |
AAG,AGG |
K390R |
XP_005278167.1 |
XM_005278111.1 |
1752 |
Missense Mutation |
AAG,AGG |
K390R |
XP_005278168.1 |
XM_005278113.1 |
1752 |
Missense Mutation |
AAG,AGG |
K390R |
XP_005278170.1 |
XM_005278114.1 |
1752 |
Missense Mutation |
AAG,AGG |
K390R |
XP_005278171.1 |
XM_005278115.1 |
1752 |
Missense Mutation |
AAG,AGG |
K390R |
XP_005278172.1 |
XM_005278116.1 |
1752 |
Missense Mutation |
AAG,AGG |
K390R |
XP_005278173.1 |
XM_005278117.1 |
1752 |
Missense Mutation |
AAG,AGG |
K390R |
XP_005278174.1 |
XM_011531071.1 |
1752 |
Missense Mutation |
AAG,AGG |
K390R |
XP_011529373.1 |
XM_011531072.1 |
1752 |
Missense Mutation |
AAG,AGG |
K390R |
XP_011529374.1 |
XM_017029987.1 |
1752 |
Missense Mutation |
AAG,AGG |
K390R |
XP_016885476.1 |
XM_017029988.1 |
1752 |
Missense Mutation |
AAG,AGG |
K390R |
XP_016885477.1 |
XM_017029989.1 |
1752 |
Missense Mutation |
AAG,AGG |
K390R |
XP_016885478.1 |
XM_017029990.1 |
1752 |
Missense Mutation |
AAG,AGG |
K390R |
XP_016885479.1 |
XM_017029991.1 |
1752 |
Missense Mutation |
AAG,AGG |
K390R |
XP_016885480.1 |
XM_017029992.1 |
1752 |
Missense Mutation |
AAG,AGG |
K390R |
XP_016885481.1 |
XM_017029993.1 |
1752 |
Missense Mutation |
AAG,AGG |
K390R |
XP_016885482.1 |
XM_017029994.1 |
1752 |
Missense Mutation |
AAG,AGG |
K390R |
XP_016885483.1 |
XM_017029995.1 |
1752 |
Missense Mutation |
AAG,AGG |
K390R |
XP_016885484.1 |
XM_017029996.1 |
1752 |
Missense Mutation |
AAG,AGG |
K390R |
XP_016885485.1 |
XM_017029997.1 |
1752 |
Missense Mutation |
AAG,AGG |
K390R |
XP_016885486.1 |
View Full Product Details