Product Details

SNP ID
rs186622147
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:3778472 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCCTGCTCCTTTTCAGCGATCTG[A/G]AGGACGAAGCTGGGGTCGCCCTGCA
Phenotype
MIM: 116949 MIM: 117140 MIM: 610674
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDC25B PubMed Links

Gene Details

Gene
CDC25B
Gene Name
cell division cycle 25B
There are no transcripts associated with this gene.

Gene
CENPB
Gene Name
centromere protein B
There are no transcripts associated with this gene.

Gene
SPEF1
Gene Name
sperm flagellar 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015417.4 679 Silent Mutation NP_056232.2
XM_005260683.4 679 Missense Mutation XP_005260740.1

View Full Product Details