Product Details

SNP ID
rs187379934
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:2299544 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTAAGTCCAAGTCCCATCCCAGC[C/T]GGATAGCCAGGGGCAACTGCCCAGG
Phenotype
MIM: 611033 MIM: 603853
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C11orf21 PubMed Links

Gene Details

Gene
C11orf21
Gene Name
chromosome 11 open reading frame 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011520034.1 65 Missense Mutation CAG,CGG Q150R XP_011518336.1
Gene
TSPAN32
Gene Name
tetraspanin 32
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139022.2 65 Intron NP_620591.3
XM_005252719.4 65 Intron XP_005252776.1
XM_005252720.4 65 Intron XP_005252777.1
XM_011519815.2 65 Intron XP_011518117.2
XM_011519818.2 65 Intron XP_011518120.2
XM_011519819.1 65 Intron XP_011518121.1
XM_011519822.1 65 Intron XP_011518124.1
XM_011519828.2 65 Intron XP_011518130.1
XM_011519829.2 65 Intron XP_011518131.2
XM_017017064.1 65 Intron XP_016872553.1
XM_017017065.1 65 Intron XP_016872554.1
XM_017017066.1 65 Intron XP_016872555.1
XM_017017067.1 65 UTR 5 XP_016872556.1
XM_017017068.1 65 Intron XP_016872557.1
XM_017017069.1 65 Intron XP_016872558.1
XM_017017070.1 65 Intron XP_016872559.1
XM_017017071.1 65 Intron XP_016872560.1

View Full Product Details